
1,1-Diisopropoxycyclohexane ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1132-95-2
:
Formula:C₁₂H₂₄O₂
MDL Number:MFCD00236381
:200.32 g/mol
Boiling Pt:85°C
EINECS:413-740-8
UN:1760
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together