kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
4,5-Difluorophthalonitrile
4,5-Difluorophthalonitrile ≥95.0% (by GC)
4,5-Difluorophthalonitrile ≥95.0% (by GC)
:
TCI
4,5-Difluorophthalonitrile ≥95.0% (by GC)
:
TCI
:
134450-56-9
:
:
C₈H₂F₂N₂
:
MFCD00061281
:
164.11 g/mol
:
3439
:
6.1,III
:
GHS06
Appearance:
White to Orange to Green powder to crystal
Purity (GC):
min. 95.0 %
Melting point:
112.0 to 116.0 deg-C
Manual Bioscience and Analyticalscience
Manual Fluorination Reagents, Fluorinated Building Blocks