Order Entry
Norway
ContactUsLinkComponent
3,5-Dimethyl-1H-pyrazole-1-carboxamide ≥98.0% (by GC, titration analysis)
3,5-Dimethyl-1H-pyrazole-1-carboxamide ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
3,5-Dimethyl-1H-pyrazole-1-carboxamide ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    110.0 to 114.0 deg-C

Specification Test Results