Order Entry
Norway
ContactUsLinkComponent
 
4,4'-Dimethoxy-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
 :  TCI
 
4,4'-Dimethoxy-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    170.0 to 174.0 deg-C