
Diphenyl-ɑ-chlorobenzylphosphonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:58263-67-5
:
Formula:C₁₉H₁₆ClO₃P
:358.76 g/mol
Boiling Pt:210°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %