
4,4'-Di-tert-butyl-2,2'-bipyridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:72914-19-3
:
Formula:C₁₈H₂₄N₂
MDL Number:MFCD01863731
:268.4000 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:159.0 to 163.0 deg-C