

4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:3945-69-5
:
Formula:C₁₀H₁₇ClN₄O₃
MDL Number:MFCD03613550
:276.72 g/mol
Storage Temperature:Freezer
EINECS:447-230-1
UN:3261
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 98.0 % (calcd.on dried substance)
- Drying loss:max. 17.0 %