
2-Chlorotrityl chloride ≥98.0% (by HPLC)
Catalog # TCIAD2504-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 95.0 %
- Melting point:135.0 to 137.0 deg-C
- Solubility in Toluene:very faint turbidity
Specification Test Results
About this item
:
:313.22 g/mol
EINECS:255-647-3
UN:3261
ADR:8,III
:GHS05
CAS Number:42074-68-0
:MFCD00278755
Formula:C₁₉H₁₄Cl₂