Order Entry
Norway
ContactUsLinkComponent
 
N,N-Diphenyl-4-methoxybenzamide ≥97.0% (by total nitrogen basis)
Catalog # TCIAD1720-5G
Supplier:  TCI
CAS Number:  
N,N-Diphenyl-4-methoxybenzamide ≥97.0% (by total nitrogen basis)
Catalog # TCIAD1720-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    140.0 to 142.0 deg-C

Specification Test Results

About this item

 :
 :303.36 g/mol
CAS Number:16034-40-5
Formula:C₂₀H₁₇NO₂
MDL Number:MFCD00059300