
Octafluoro-4,4'-biphenol ≥95.0% (by GC)
Catalog # TCIAD1519-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:209.0 to 213.0 deg-C
Specification Test Results
About this item
:
:330.13 g/mol
:GHS07
CAS Number:2200-70-6
Formula:C₁₂H₂F₈O₂
MDL Number:MFCD00045743