Order Entry
Norway
ContactUsLinkComponent
2,4-Dimethyl-1-hexene ≥98.0% (by GC)
2,4-Dimethyl-1-hexene ≥98.0% (by GC)
Catalog # TCIAD1241-5ML
Supplier:  TCI
CAS Number:  
2,4-Dimethyl-1-hexene ≥98.0% (by GC)
Catalog # TCIAD1241-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:111°C
Flash Pt:8°C
ADR:3,II
 :112.22 g/mol
UN:3295
 :GHS02
 :GHS08
CAS Number:16746-87-5
Formula:C₈H₁₆
MDL Number:MFCD00048655