β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate ≥95.0% (by HPLC) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:53-84-9
:
Formula:C₂₁H₂₇N₇O₁₄P₂·H₂O
MDL Number:MFCD00036253
Merck Index:14,06344
Storage Temperature:Refrigerator
EINECS:200-184-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- abs-ratio (a/b):0.80 to 0.86 (E250/E260)
- abs-ratio (2a/2b):0.19 to 0.23 (E280/E260)
- Molar extinction coefficient:17,000 to 18,500 (H2O, 260.0 nm) (calcd.on anh.substance)
Specification Test Results
Frequently Bought Together