Benzil dioxime ≥98.0% (by total nitrogen basis)
TCIAD0891-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Solubility in Pyridine:almost transparency
:
:240.26 g/mol
:14,01079
:245-921-0
:23873-81-6
:C₁₄H₁₂N₂O₂
:MFCD00002113