Order Entry
Norway
ContactUsLinkComponent
Diphenhydramine hydrochloride ≥98.0% (by HPLC, titration analysis)
Irritant
Diphenhydramine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
Diphenhydramine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    168.0 to 172.0 deg-C
  • Solubility in Water:
    almost transparency

Specification Test Results

Related Information