[11]Cycloparaphenylene ≥98.0% (by HPLC)
Catalog # TCIAC3536-10MG
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:837.08 g/mol
CAS Number:1222105-48-7
Formula:C₆₆H₄₄