Order Entry
Norway
ContactUsLinkComponent
Coomassie brilliant blue G-250 solution for electrophoresis, ready-to-use
Coomassie brilliant blue G-250 solution for electrophoresis, ready-to-use
Catalog # TCIAC3488-500ML
Supplier:  TCI
CAS Number:  
Coomassie brilliant blue G-250 solution for electrophoresis, ready-to-use
Catalog # TCIAC3488-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Green to Dark green to Dark blue clear liquid
  • Protein staining test:
    to pass test

Specification Test Results

About this item

 :
 :854.04 g/mol
 :GHS07
 :GHS08
CAS Number:6104-58-1
Formula:C₄₇H₄₈N₃NaO₇S₂
MDL Number:MFCD00078482