Urethane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51-79-6
:
Formula:C₃H₇NO₂
MDL Number:MFCD00007966
:89.09 g/mol
Merck Index:12,10013
Boiling Pt:182…184 °C (760 mmHg)
Melting Pt:47…51 °C
Density:1,11 g/cm³ (20 °C)
Flash Pt:92 °C
Storage Temperature:Ambient
EINECS:200-123-1
ADR:6.1,II
:GHS08
:GHS07
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:46 to 51 °C
- GC:≥96.0 %