6-Chloroimidazo[1,2-a]pyridine ≥98.0% (by GC, titration analysis)
: TCI
:6188-25-6
:
:C₇H₅ClN₂
:MFCD05663807
:152.58 g/mol
:132°C
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:85.0 to 89.0 deg-C