Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Chloromethyl)benzothiazole
2-(Chloromethyl)benzothiazole ≥98.0% (by GC)
2-(Chloromethyl)benzothiazole ≥98.0% (by GC)
TCIAC3335-1G
:
TCI
:
2-(Chloromethyl)benzothiazole ≥98.0% (by GC)
TCIAC3335-1G
:
TCI
:
Pk:
1 g
Seq:
3047
Purity (GC):
min. 98.0 %
:
:
183.65 g/mol
:
90°C
:
GHS05
:
GHS07
:
Refrigerator
:
37859-43-1
:
C₈H₆CINS
:
MFCD05664944