2-Chloro-N-(2-chloro-4-methyl-3-pyridyl)nicotinamide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:133627-46-0
:
Formula:C₁₂H₉Cl₂N₃O
MDL Number:MFCD09031256
:282.12 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:193.0 to 197.0 deg-C