
3-Cyano-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAC3214-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):98.0 to 112.0 %
Specification Test Results
About this item
:
:164.9295 g/mol
:GHS07
CAS Number:957121-05-0
Formula:C₇H₅BFNO₂
MDL Number:MFCD07374984