Order Entry
Norway
ContactUsLinkComponent
2-(3'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(3'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
2-(3'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    192.0 to 196.0 deg-C

Specification Test Results