Order Entry
Norway
ContactUsLinkComponent
4-Cyano-4'-hexylbiphenyl ≥98.0% (by GC)
Irritant
Environment
4-Cyano-4'-hexylbiphenyl ≥98.0% (by GC)
Supplier:  TCI
Certificates
Irritant
Environment
4-Cyano-4'-hexylbiphenyl ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid to cloudy liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results