


1-Chloro-2,6-difluoro-3-nitrobenzene ≥98.0% (by GC)
Catalog # TCIAC2964-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:45.0 to 49.0 deg-C
Specification Test Results
About this item
:
:193.54 g/mol
EINECS:411-980-8
UN:1759
ADR:8,II
:GHS05
:GHS07
:GHS09
CAS Number:3847-58-3
Formula:C₆H₂ClF₂NO₂
MDL Number:MFCD00042201