
2-Chloroimidazole ≥98.0% (by GC)
Catalog # TCIAC2685-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:166.0 to 170.0 deg-C
Specification Test Results
About this item
:
:102.52 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:16265-04-6
Formula:C₃H₃ClN₂
MDL Number:MFCD02179530