

4-Chlorophenyl chloroformate ≥98.0% (by GC, titration analysis)
: TCI
:7693-45-0
:
:C₇H₄Cl₂O₂
:MFCD00000638
:102°C
:103°C
:3277
:6.1,II
:GHS05
:GHS06
:Refrigerator
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %