5'-Chloro-3-hydroxy-2'-methyl-2-naphthanilide ≥96.0% (by total nitrogen basis)
Catalog # TCIAC2102-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (with Total Nitrogen):min. 96.0 %
Specification Test Results
About this item
:
:311.77 g/mol
Melting Pt:244°C
EINECS:205-207-1
CAS Number:135-63-7
Formula:C₁₈H₁₄ClNO₂
MDL Number:MFCD00021636