
4-Cyano-2,2,6,6-tetramethylpiperidine-1-oxyl ≥95.0% (by titrimetric analysis) free radical
Catalog # TCIAC1782-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 93.0 %
- Purity (Iodometric Titration):min. 95.0 %
- Melting point:143.0 to 149.0 deg-C
Specification Test Results
About this item
:
:181.26 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:38078-71-6
Formula:C₁₀H₁₇N₂O
MDL Number:MFCD00191964