Order Entry
Norway
ContactUsLinkComponent
2-Chloroquinoline-4-carboxylic acid ≥97.0% (by HPLC, titration analysis)
Irritant
2-Chloroquinoline-4-carboxylic acid ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
2-Chloroquinoline-4-carboxylic acid ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 97.0 %

Specification Test Results

Frequently Bought Together