
2-Chloro-3-methoxypropionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2544-05-0
:
Formula:C₄H₇ClO₃
MDL Number:MFCD03093624
:138.55 g/mol
Boiling Pt:155°C
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %