

(3-Chloropropyl)diphenylsulphonium tetrafluoroborate ≥98.0% (by HPLC)
Catalog # TCIAC1380-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:107.0 to 111.0 deg-C
Specification Test Results
About this item
:
:350.61 g/mol
UN:1759
ADR:8,III
:GHS05
:GHS07
CAS Number:33462-80-5
Formula:C₁₅H₁₆BClF₄S
MDL Number:MFCD00074932