

Chlorazol black E
Catalog # TCIAC0533-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Black powder to crystal
- Elemental analysis (Nitrogen):min. 8.00 %
Specification Test Results
About this item
:
:781.73 g/mol
EINECS:217-710-3
:GHS07
:GHS08
CAS Number:1937-37-7
Formula:C₃₄H₂₇N₉O₇S₂
MDL Number:MFCD00066363