Order Entry
Norway
ContactUsLinkComponent
3'-Chloroindophenol sodium salt redox indicator
Irritant
3'-Chloroindophenol sodium salt redox indicator
Catalog # TCIAC0188-1G
Supplier:  TCI
CAS Number:  
Irritant
3'-Chloroindophenol sodium salt redox indicator
Catalog # TCIAC0188-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Dark green to Dark blue to Black powder to crystal
  • Sensitiveness:
    to pass test

Specification Test Results

About this item

 :
 :255.63 g/mol
 :GHS07
CAS Number:41350-02-1
Formula:C₁₂H₈ClNO₂
MDL Number:MFCD00059876