(Boc-amino)-PEG3-C2-Carboxylic acid ≥97.0% (by HPLC, titration analysis)
TCIAB6093-250MG
: TCI
:
- Pk:250 mg
- Seq:2590
- Appearance:Colorless to Light yellow clear liquid
- Purity (HPLC) (CAD):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0 %
:
:321.37 g/mol
:Freezer
:1347750-75-7
:C₁₄H₂₇NO₇
:MFCD24539476