Order Entry
Norway
ContactUsLinkComponent
 
2-(4-Bromophenyl)-2-methylpropanenitrile ≥95.0% (by GC)
 :  TCI
 
2-(4-Bromophenyl)-2-methylpropanenitrile ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %