
2-Bromo-5-methoxypyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:105170-27-2
:
Formula:C₆H₆BrNO
MDL Number:MFCD07781221
:188.02 g/mol
Boiling Pt:78°C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %