
2-Bromo-5-methylbenzaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:90221-55-9
:
Formula:C₈H₇BrO
MDL Number:MFCD08669550
:199.05 g/mol
Boiling Pt:100°C
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:41.0 to 45.0 deg-C