4-Bromobenzaldehyde dimethyl acetal ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:24856-58-4
:
Formula:C₉H₁₁BrO₂
MDL Number:MFCD01320499
:231.09 g/mol
Boiling Pt:94°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %