9-Bromo-10-[3-(2-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:944801-33-6
:
Formula:C₃₀H₁₉Br
MDL Number:MFCD30470616
:459.39 g/mol
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 95.0 %
- Melting point:193.0 to 197.0 deg-C