Order Entry
Norway
ContactUsLinkComponent
9-Bromo-10-[3-(1-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)
9-Bromo-10-[3-(1-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)
Catalog # TCIAB5771-1G
Supplier:  TCI
CAS Number:  
9-Bromo-10-[3-(1-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)
Catalog # TCIAB5771-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    177.0 to 181.0 deg-C

Specification Test Results

About this item

 :
 :459.39 g/mol
CAS Number:1304129-94-9
Formula:C₃₀H₁₉Br
MDL Number:MFCD28291941