Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
SB216763
SB216763 ≥98.0% (by HPLC)
SB216763 ≥98.0% (by HPLC)
:
TCI
SB216763 ≥98.0% (by HPLC)
:
TCI
:
280744-09-4
:
:
C₁₉H₁₂Cl₂N₂O₂
:
MFCD09753369
:
371.2 g/mol
:
104°C
:
Freezer
Appearance:
Light yellow to Brown powder to crystal
Purity (HPLC):
min. 98.0 area%