N,N-Bis(4'-diphenylamino-4-biphenylyl)amine ≥95.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:167218-39-5
:
Formula:C₄₈H₃₇N₃
MDL Number:MFCD32263460
:655.85 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
- Melting point:209.0 to 213.0 deg-C