Order Entry
Norway
ContactUsLinkComponent
1,4-Bis(trifluoromethyl)cyclohexane cis- and trans-mixture ≥98.0% (by GC)
Flammable
1,4-Bis(trifluoromethyl)cyclohexane cis- and trans-mixture ≥98.0% (by GC)
Supplier:  TCI
Certificates
Flammable
1,4-Bis(trifluoromethyl)cyclohexane cis- and trans-mixture ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomer)

Specification Test Results