kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
2-Bromo-9,9-dimethyl-9,10-dihydroacridine
2-Bromo-9,9-dimethyl-9,10-dihydroacridine ≥96.0% (by GC)
2-Bromo-9,9-dimethyl-9,10-dihydroacridine ≥96.0% (by GC)
:
TCI
2-Bromo-9,9-dimethyl-9,10-dihydroacridine ≥96.0% (by GC)
:
TCI
:
1443680-94-1
:
:
C₁₅H₁₄BrN
:
288.19 g/mol
Appearance:
White to Orange to Green powder to crystal
Purity (GC):
min. 96.0 %
Melting point:
95.0 to 99.0 deg-C