Order Entry
Norway
ContactUsLinkComponent
 
[3-(Bromomethyl)oxetan-3-yl]methanol ≥97.0% (by GC)
  TCIAB5442-1G
 :  TCI
 :  
[3-(Bromomethyl)oxetan-3-yl]methanol ≥97.0% (by GC)
  TCIAB5442-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :181.04 g/mol
 :150°C
 :100°C
 :22633-44-9
 :C₅H₉BrO₂
 :MFCD09800598