Nalpha-tert-Butoxycarbonyl-Nepsilon-tetradecanoyl-L-lysine ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2173052-71-4
:
Formula:C₂₅H₄₈N₂O₅
MDL Number:MFCD30725738
:456.67 g/mol
Specification Test Results
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:87.0 to 92.0 deg-C