Order Entry
Norway
ContactUsLinkComponent
2,2'-Bithiophene-5,5'-dicarboxaldehyde ≥98.0% (by GC)
Irritant
2,2'-Bithiophene-5,5'-dicarboxaldehyde ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
2,2'-Bithiophene-5,5'-dicarboxaldehyde ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Yellow to Amber powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Frequently Bought Together