
5-Bromo-2-chloro-4-fluoroanisole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:146447-18-9
:
Formula:C₇H₅BrClFO
MDL Number:MFCD07784265
:239.47 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:72.0 to 76.0 deg-C