
Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:95715-86-9
:
Formula:C₁₂H₂₁NO₅
MDL Number:MFCD00674041
:259.302 g/mol
Boiling Pt:102°C
Flash Pt:93°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 96.0 %
- Optical purity (GC):min. 96.0 ee%