Order Entry
Norway
ContactUsLinkComponent
 
 
5'-Bromo-2'-fluoroacetophenone ≥98.0% (by GC)
 :  TCI
 
 
5'-Bromo-2'-fluoroacetophenone ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    40.0 to 44.0 deg-C

 

Frequently Bought Together